Learn More
Abcam Spectinomycin hydrochloride pentahydrate, aminocylcitol antibiotic, 5G
Catalog No. 502706387
Encompass_Preferred
MW 368.81 Da, Purity >98%. An aminocyclcitol antibiotic. Active against Gram-positive and Gram-negative bacteria. Binds the 30S ribosome subunit and interrupts protein synthesis by interfering with peptidyl tRNA translocation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6387 Supplier Abcam Supplier No. AB1419685G
MW 368.81 Da, Purity >98%. An aminocyclcitol antibiotic. Active against Gram-positive and Gram-negative bacteria. Binds the 30S ribosome subunit and interrupts protein synthesis by interfering with peptidyl tRNA translocation.
The product is subject to the following: Abcam Restricted Use Statement