Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111004756 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4756 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4756 Supplier MilliporeSigma Supelco Supplier No. PHR1426500MG
Add to Cart
Add to Cart
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Spectinomycin Hydrochloride is an aminocyclitol antibiotic, widely used in human medicine for the treatment of uncomplicated gonorrhea and also in veterinary medicine to treat bacterial respiratory and enteric infections. Its mode of action involves the inhibition of protein synthesis in the bacterial cell wall.
TRUSTED_SUSTAINABILITY

Specifications

Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
MDL Number MFCD04041129
Synonym Spectinomycin
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.