Learn More
Alkali Scientific Spectinomycin Sulfate, 25 G
SDP
Catalog No. 5000660982
Encompass_Preferred
Spectinomycin Sulfate, 25g, is a potent antibiotic available in powder form, used primarily to treat infections caused by Neisseria gonorrhoeae. The 25g quantity makes it ideal for research laboratories and clinical settings that require larger quantities for continuous or bulk treatment. Spectinomycin inhibits bacterial protein synthesis by targeting the 30S ribosomal subunit, effectively halting bacterial growth. It is especially effective in treating penicillin-resistant strains of Neisseria gonorrhoeae, making it a crucial option in managing sexually transmitted infections. Additionally, Spectinomycin Sulfate is used in research for studying antibiotic resistance and bacterial translation.
Catalog No. 50-006-60982 Supplier Alkali Scientific Supplier No. PEC002CP
Spectinomycin Sulfate, 25g, is a potent antibiotic available in powder form, used primarily to treat infections caused by Neisseria gonorrhoeae. The 25g quantity makes it ideal for research laboratories and clinical settings that require larger quantities for continuous or bulk treatment. Spectinomycin inhibits bacterial protein synthesis by targeting the 30S ribosomal subunit, effectively halting bacterial growth. It is especially effective in treating penicillin-resistant strains of Neisseria gonorrhoeae, making it a crucial option in managing sexually transmitted infections. Additionally, Spectinomycin Sulfate is used in research for studying antibiotic resistance and bacterial translation.