Learn More
Electron Microscopy Sciences SpecTainer™ II Container Conventional, 120 mL, Sterile Sterile, Cyan
SDP
Catalog No. 501931650
Encompass_Preferred
This container uses a conventional leakproof screw cap with molded ridges around the lid making it easy to open and close. It is designed for collection, transport, and storage of specimens. The polypropylene containers are safe to use even in adverse conditions. Containers are uniquely stackable, shatter resistant and are manufactured from virgin high-clarity polypropylene. They are designed with a straight side format. The warning label has space for patient identification. Graduated.
Volume: 120 mL
Sterile: Sterile
Catalog No. 50-193-1650 Supplier Electron Microscopy Sciences Supplier No. 6424924
This container uses a conventional leakproof screw cap with molded ridges around the lid making it easy to open and close. It is designed for collection, transport, and storage of specimens. The polypropylene containers are safe to use even in adverse conditions. Containers are uniquely stackable, shatter resistant and are manufactured from virgin high-clarity polypropylene. They are designed with a straight side format. The warning label has space for patient identification. Graduated.
Volume: 120 mL
Sterile: Sterile