Learn More
Electron Microscopy Sciences SpecTainer™ I Container Tamper Evident, 120 mL, Yellow
SDP
Catalog No. 501931641
Encompass_Preferred
The Spectainer I is incorporates a unique tamper evident screw cap, ensuring sample integrety and peace of mind during transport or storage situations where someone could tamper with the specimen. The molded ridges around lid make it easy to open and close. The warning label has space for patient identification. Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant.
Volume: 120 mL
Color: Yellow
Catalog No. 50-193-1641 Supplier Electron Microscopy Sciences Supplier No. 6424915
The Spectainer I is incorporates a unique tamper evident screw cap, ensuring sample integrety and peace of mind during transport or storage situations where someone could tamper with the specimen. The molded ridges around lid make it easy to open and close. The warning label has space for patient identification. Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant.
Volume: 120 mL
Color: Yellow