Learn More
Electron Microscopy Sciences SpecTainer™ I Container Tamper Evident, 60 mL, Sterile
SDP
Catalog No. 501931642
Encompass_Preferred
The Spectainer I is incorporates a unique tamper evident screw cap, ensuring sample integrety and peace of mind during transport or storage situations where someone could tamper with the specimen. The molded ridges around lid make it easy to open and close. The warning label has space for patient identification. Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant.
Volume: 60 mL
Sterile: Sterile
Color: Cyan
Catalog No. 50-193-1642 Supplier Electron Microscopy Sciences Supplier No. 6424916
Add to Cart
Add to Cart
The Spectainer I is incorporates a unique tamper evident screw cap, ensuring sample integrety and peace of mind during transport or storage situations where someone could tamper with the specimen. The molded ridges around lid make it easy to open and close. The warning label has space for patient identification. Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant.
Volume: 60 mL
Sterile: Sterile
Color: Cyan