Learn More
Electron Microscopy Sciences Eco SpecTainer™ Container Tamper Evident, 60 mL, Gold
SDP
Catalog No. 501931627
Encompass_Preferred
These eco-friendly, rapidly degradable polypropylene containers are one of the most innovative specimen containers on the market today. Plastics take hundreds of years to degrade naturally in the environment, while the only condition necessary for these to biodegrade is constant contact with other degrading material.
No heat, physical stress, oxygen or sunlight are needed for product breakdown. They have the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment. They will biodegrade to become some of the soil's organic components in less than 7 years in constant contact with the soil, instead of up to 400 years as conventional plastics.
Volume: 60 mL
Catalog No. 50-193-1627 Supplier Electron Microscopy Sciences Supplier No. 6424901
These eco-friendly, rapidly degradable polypropylene containers are one of the most innovative specimen containers on the market today. Plastics take hundreds of years to degrade naturally in the environment, while the only condition necessary for these to biodegrade is constant contact with other degrading material.
No heat, physical stress, oxygen or sunlight are needed for product breakdown. They have the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment. They will biodegrade to become some of the soil's organic components in less than 7 years in constant contact with the soil, instead of up to 400 years as conventional plastics.
Volume: 60 mL