Learn More
Leica Biosystems SPECTRA CV X1 MOUNTING MEDIUM
Catalog No. NC1589769
Encompass_Preferred
HistoCore SPECTRA CV X1 is a rapid drying, xylene based mounting media. An antioxidant is added to inhibit stain fading and prevent the formation of annual rings. It sets up quickly, allowing slide preparations to be handled immediately after mounting. SPECTRA X1 is soluble in xylene, toluene, and most xylene substitutes.This product is only available for use in conjunction with the HistoCore SPECTRA CV.
Catalog No. NC1589769 Supplier Leica Biosystems Supplier No. 3801733
HistoCore SPECTRA CV X1 is a rapid drying, xylene based mounting media. An antioxidant is added to inhibit stain fading and prevent the formation of annual rings. It sets up quickly, allowing slide preparations to be handled immediately after mounting. SPECTRA X1 is soluble in xylene, toluene, and most xylene substitutes.This product is only available for use in conjunction with the HistoCore SPECTRA CV.