Learn More
Description
Obtain faster results with no need for reincubation of negatives with Thermo Scientific™ Spectra™ MRSA, a new, improved chromogenic screening plate for Methicillin Resistant Staphylococcus aureus (MRSA). The medium combines excellent sensitivity with the simplicity of a chromogenic plate to create an invaluable tool in the implementation of infection control policies and procedures.
- Rapidly and economically detects nasal colonization of MRSA and MRSA from positive blood cultures
- Aids in the prevention of MRSA infections
- Delivers high Positive Predictive Value (PPV) to reduce unnecessary costs
- Enables quick, efficient screening of patients
- Produces easy-to-read, denim blue colonies on an opaque background
- Requires no special handling for optimal workflow 24/7
Spectra MRSA is a selective medium for the screening of clinical samples for the presence of Methicillin-Resistant Staphylococcus aureus (MRSA), with results available within 18 hours and no need to reincubate negatives. The opaque medium incorporates a novel chromogen that yields a blue color as a result of phosphatase activity. This enzyme is present in many staphylococci including Staphylococcus aureus. To allow the differentiation of MRSA accurately, the medium contains a combination of antibacterial compounds designed to inhibit the growth of a wide variety of competitor organisms and MSSA. Also included are compounds to suppress the expression of phosphatase activity in other staphylococci, thus ensuring a high level of sensitivity and specificity.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Opaque light tan |
| Content And Storage | 2°C to 8°C |
| Format | 85mm Monoplate |
| Manufacturing Origin | United States |
| Media Type | Spectra MRSA Medium |
| Product Type | Agar Plate |
| Fill Volume | 18 mL |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
