Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Spectra™ MRSA Medium
GSA_VA

Catalog No. r01821
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01821 10/Pk.
R01822 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01821 Supplier Thermo Scientific™ Supplier No. R01821
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus.

Obtain faster results with no need for reincubation of negatives with Thermo Scientific™ Spectra™ MRSA, a new, improved chromogenic screening plate for Methicillin Resistant Staphylococcus aureus (MRSA). The medium combines excellent sensitivity with the simplicity of a chromogenic plate to create an invaluable tool in the implementation of infection control policies and procedures.

Limit the spread of MRSA by incorporating an effective screening protocol as part of your infection control program. Colonized patients must be targeted for isolation and appropriate treatment as early as possible; therefore, speed and accuracy of results are recognized as critical. Save time and infection control resources by giving negative patients the all-clear faster and more reliably.
  • Rapidly and economically detects nasal colonization of MRSA and MRSA from positive blood cultures
  • Aids in the prevention of MRSA infections
  • Delivers high Positive Predictive Value (PPV) to reduce unnecessary costs
  • Enables quick, efficient screening of patients
  • Produces easy-to-read, denim blue colonies on an opaque background
  • Requires no special handling for optimal workflow 24/7

Spectra MRSA is a selective medium for the screening of clinical samples for the presence of Methicillin-Resistant Staphylococcus aureus (MRSA), with results available within 18 hours and no need to reincubate negatives. The opaque medium incorporates a novel chromogen that yields a blue color as a result of phosphatase activity. This enzyme is present in many staphylococci including Staphylococcus aureus. To allow the differentiation of MRSA accurately, the medium contains a combination of antibacterial compounds designed to inhibit the growth of a wide variety of competitor organisms and MSSA. Also included are compounds to suppress the expression of phosphatase activity in other staphylococci, thus ensuring a high level of sensitivity and specificity.


Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque light tan
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type Spectra MRSA Medium
Product Type Agar Plate
Fill Volume 18 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.