Learn More
Crescent Chemical Co Inc SPECTRA/POR 1 DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44171.01
Encompass_Preferred
SPECTRA/POR 1 dialysis tubing, MWCO 6000 - 8000 RC, diameter 14.6 mm Packed dry, with glycerol as protection for embrittlement, which can easily be removed by soaking in water. Contains low level of heavy metal and sulfide impurities. Size - 30M Storage Conditions - +15 C TO +30 C
Catalog No. 50-248-878