Learn More
Crescent Chemical Co Inc SPECTRA/POR 1 DIALYSIS TUBING
SDP
Catalog No. 50248880
Encompass_Preferred
SPECTRA/POR 1 dialysis tubing, MWCO 6000 - 8000 RC, diameter 32 mm Packed dry, with glycerol as protection for embrittlement, which can easily be removed by soaking in water. Contains low level of heavy metal and sulfide impurities. Size - 30M Storage Conditions - +15 C TO +30 C /
Catalog No. 50-248-880 Supplier Crescent Chemical Co Inc Supplier No. SE44174.01
SPECTRA/POR 1 dialysis tubing, MWCO 6000 - 8000 RC, diameter 32 mm Packed dry, with glycerol as protection for embrittlement, which can easily be removed by soaking in water. Contains low level of heavy metal and sulfide impurities. Size - 30M Storage Conditions - +15 C TO +30 C /