Learn More
Crescent Chemical Co Inc SPECTRA/POR 3 DIALYSIS TUBING
SDP
Catalog No. 50248883
Encompass_Preferred
SPECTRA/POR 3 dialysis tubing, MWCO 3500 RC, diameter 11.5 mm Packed dry, with glycerol as protection for embrittlement, which can easily be removed by soaking in water. Contains low level of heavy metal and sulfide impurities. Size - 15M Storage Conditions - +15 C TO +30 C
Catalog No. 50-248-883 Supplier Crescent Chemical Co Inc Supplier No. SE44183.01
SPECTRA/POR 3 dialysis tubing, MWCO 3500 RC, diameter 11.5 mm Packed dry, with glycerol as protection for embrittlement, which can easily be removed by soaking in water. Contains low level of heavy metal and sulfide impurities. Size - 15M Storage Conditions - +15 C TO +30 C