Learn More
Crescent Chemical Co Inc SPECTRA/POR 6 DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44205.01
Encompass_Preferred
SPECTRA/POR 6 dialysis tubing, MWCO 8000 RC, diameter 15 mm Pre-wetted (containing 0.1 % sodium azide). Readyto-use, no soaking for removal of glycerol necessary. Contains low level of heavy metal and sulfide impurities. Size - 10M Storage Conditions - +15 C TO +30 C Ca
Catalog No. 50-248-898