Learn More
Crescent Chemical Co Inc SPECTRA/POR 6 DIALYSIS TUBING
SDP
Catalog No. 50248897
Encompass_Preferred
SPECTRA/POR 6 dialysis tubing, MWCO 8000 RC, diameter 11.5 mm Pre-wetted (containing 0.05 % sodium azide). Readyto-use, no soaking for removal of glycerol necessary. Contains low level of heavy metal and sulfide impurities. Size - 10M Storage Conditions - +15 C TO +30 C
Catalog No. 50-248-897 Supplier Crescent Chemical Co Inc Supplier No. SE44204.01
SPECTRA/POR 6 dialysis tubing, MWCO 8000 RC, diameter 11.5 mm Pre-wetted (containing 0.05 % sodium azide). Readyto-use, no soaking for removal of glycerol necessary. Contains low level of heavy metal and sulfide impurities. Size - 10M Storage Conditions - +15 C TO +30 C