Learn More
Crescent Chemical Co Inc SPECTRA/POR 6 DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44228.01
Encompass_Preferred
SPECTRA/POR 6 dialysis tubing, MWCO 50 000 RC, diameter 7.5 mm Pre-wetted (containing 0.05 % sodium azide). Readyto-use, no soaking for removal of glycerol necessary. Contains low level of heavy metal and sulfide impurities. Size - 10M Storage Conditions - +15 C TO +30 C
Catalog No. 50-248-921