Learn More
Thermco Products Inc -40 deg./25 deg.C & -40 deg./80 deg.F Refrigerator/Freezer Spirit
Catalog No. NC0107952
Encompass_Preferred
Ideal for both Refrigerators and Freezers, our dual-scale thermometer has a range of -40 to 25°C and -40 to 80°F. It features an enclosed bulb to help ensure average temperature readings and an open scale for easy, accurate readings. The thermometer’s rust-resistant case has stainless steel hangers with keyhole brackets for easy hanging and removal. Enamelled aluminium scale. Thermometer can hang or stand.
Catalog No. NC0107952 Supplier Thermco Products Inc Supplier No. BFZ80
Ideal for both Refrigerators and Freezers, our dual-scale thermometer has a range of -40 to 25°C and -40 to 80°F. It features an enclosed bulb to help ensure average temperature readings and an open scale for easy, accurate readings. The thermometer’s rust-resistant case has stainless steel hangers with keyhole brackets for easy hanging and removal. Enamelled aluminium scale. Thermometer can hang or stand.