Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Spectra™ UTI Medium

Catalog No. 23246901 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-246-901 100/Pk.
23-246-900 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-246-901 Supplier Thermo Scientific™ Supplier No. R01819
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain fast and accurate results with a chromogenic medium designed for screening clinical samples for the presence of organisms commonly associated with urinary pathogens.

Isolate, enumerate and presumptively identify common organisms associated with urinary tract infections within 16 to 24 hours using Thermo Scientific™ Spectra™ UTI. Organisms are differentiated through the utilization of one or more chromogens and/or Tryptophan. Interpretation is aided by the inclusion of an opaque background, providing a distinct contrast against the seven diagnostic colors of colony growth.

The UTI formulation contains two chromogenic substrates which are cleaved by the ß-galactosidase and ß-glucosidase enzymes produced by E. coli, Enterococcus species and coliforms. These specific enzyme reactions cleave the chromogens giving a range of diagnostic colors. The ß-galactosidase activity of E. coli and Staph. saprophyticus results in pink/red colonies, and the ß-glucosidase activity of enterococci produces blue/turquoise colonies; the activity of both enzymes on coliforms gives dark blue/purple colonies. Tryptophan deaminase activity produces a brown halo around colonies of Proteus, Morganella and Providencia species. Most other organisms exhibit their natural pigmentation.

The specificity of Brilliance UTI minimizes confirmatory testing with same day results for E.coli confirmation being achieved using a rapid indole test (DMAC) (MB1448A).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Opaque light tan
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type Spectra UTI Medium
Product Type Agar Plate
Fill Volume 18 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.