Description
Thermo Scientific™ Remel Spectra UTI/Sheep Blood is a chromogenic medium for isolation, differentiation and presumptive identification of pathogenic microorganisms associated with urinary tract infections.
Specifications
Specifications
| Description | Spectra UTI // Sheep Blood |
| Format | Biplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |