Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Spectra™ VRE Medium
GSA_VA

Catalog No. r01830
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
85mm Monoplate
Media Type:
Spectra VRE Medium
Manufacturing Origin:
United States
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R01830 10/Pk. 85mm Monoplate Spectra VRE Medium United States
R01832 100/Pk. 85mm Monoplate Spectra VRE Medium United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01830 Supplier Thermo Scientific™ Supplier No. R01830
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain fast and accurate results with a chromogenic medium designed for screening clinical samples for the presence of vancomycin-resistant Enterococcus (VRE).

Improve the prevention and control of VRE in healthcare settings by rapidly and reliably isolating and presumptively identifying vancomycin-resistant Enterococcus (VRE) within 24 hours, direct from sample, using Thermo Scientific™ Spectra™ VRE. Vancomycin-resistant Enterococcus faecium and Enterococcus faecalis are differentiated through the utilization of two separate chromogens. Interpretation is aided through the inclusion of a semi-opaque background, providing a distinct visual contrast against light blue and indigo colonies.

Differentiation of vancomycin resistant E. faecium from E. faecalis is achieved through the inclusion of two chromogens that are targeted by specific enzymes: phosphatase and α-galactosidase. The action of these enzymes on the chromogens results in a build-up of color within the colony. The color produced depends on which enzymes the organisms possess. The presence of phosphatase enzymes in both E. faecium and E. faecalis results in a light blue colony; however, E. faecium also produces α-galactosidase resulting in a mix of blue and pink chromophores within the bacterium, producing indigo to purple colonies which are easily distinguished from the light blue E. faecalis colonies.

Additional antibiotics, in combination with vancomycin, are present to suppress the growth of competing flora including E. gallinarum and E. casseliflavus, both of which are intrinsically resistant to vancomycin, possessing the chromosomally encoded VanC resistance mechanism. The VanC resistance mechanism is not readily transmissible between organisms and, as such, is deemed less clinically significant than VanA and VanB mechanisms which are encoded on freely transmissible genetic elements, plasmids and transposons, thus increasing the risk of resistance genes spreading to other organisms.
  • Rapidly and economically detects colonization of Vancomycin-Resistant Enterococcus (VRE) colonization
  • Aids in the prevention and controls the spread of vancomycin resistance in healthcare settings
  • Correctly classifies patients whether positive or negative
  • Provides superior results in a selective and differential chromogenic medium

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Semi-opaque light tan
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type Spectra VRE Medium
Product Type Agar
Fill Volume 18 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.