Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES SpectraComp, Single vial (pos+neg combined), 100 tests
SDP

Catalog No. NC2096619
Encompass_Preferred

SpectraComp controls are precision engineered single stain cell mimics designed to deliver accurate spectral unmixing and compensation by matching the staining performance and autofluorescence of real cells.

Specifications:
Catalog Number: SSB-05-B
Format: Single vial (pos+neg combined)
Package Size: 100 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 18 months
Country of Origin: United States
For Research Use Only.

This item is not returnable. View return policy
Catalog No. NC2096619 Supplier SLINGSHOT BIOSCIENCES Supplier No. SSB05B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SpectraComp controls are precision engineered single stain cell mimics designed to deliver accurate spectral unmixing and compensation by matching the staining performance and autofluorescence of real cells.

Specifications:
Catalog Number: SSB-05-B
Format: Single vial (pos+neg combined)
Package Size: 100 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 18 months
Country of Origin: United States
For Research Use Only.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.