Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES SpectraComp eGFP, 4 vials (neg/low/med/high), 25 tests
SDP

Catalog No. NC2950588
Encompass_Preferred

SpectraComp eGFP controls are precision engineered multi level fluorescent cell mimics designed for accurate spectral unmixing, compensation and benchmarking of eGFP expressing cells in spectral and conventional flow cytometry workflows.

Specifications:
Catalog Number: SSB-23-A
Format: 4 vials (neg/low/med/high)
Package Size: 25 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 24 months
Country of Origin: United States
For Research Use Only.

Catalog No. NC2950588 Supplier SLINGSHOT BIOSCIENCES Supplier No. SSB23A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SpectraComp eGFP controls are precision engineered multi level fluorescent cell mimics designed for accurate spectral unmixing, compensation and benchmarking of eGFP expressing cells in spectral and conventional flow cytometry workflows.

Specifications:
Catalog Number: SSB-23-A
Format: 4 vials (neg/low/med/high)
Package Size: 25 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 24 months
Country of Origin: United States
For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.