Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD™ SpectraComp™ Unmixing and Compensation Particles 100 Tests RUO

Catalog No. BDB569991 Shop All BD Biosciences Products
Change view
Click to view available options
For Use With (Equipment):
Flow Cytometers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
BDB569991 Flow Cytometers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569991 Supplier BD™ Supplier No. 569991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A multi-species reference control, used for correcting fluorescence spillover in both conventional and spectral flow cytometry

  • Simplify setting up controls for experiments as they expand the range of antibodies that can be recognized with a single bead.
  • Microparticles used to help generate fluorochrome profiles for spectral unmixing and compensation in multicolor flow cytometry.
  • Contains both a positive and negative particle in the same dropper bottle.
  • The positive particle can bind both kappa and lambda light chain-bearing immunoglobulins raised in mouse, rat, and hamster hosts.
  • Low autofluorescence on all laser lines, including violet and UV.
  • Compatible with common used stain, lysis, and fix/perm buffers.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Application) Flow Cytometry
Product Type Compensation Beads
For Use With (Equipment) Flow Cytometers
No. of Tests 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.