Learn More
Molecular Devices Molecular Devices Micro-Volume Microplate Kit, HTS, 24-Well and 64-Well, 2 μL/4 μL
Catalog No. 5000014374
Encompass_Preferred
The SpectraDrop High-Throughput Screening (HTS) Kit provides consumables and a microplate adapter to enable automated, low-volume sample measurement using micro-volume microplates. It combines multiple low-volume slide formats and cover slides to support high-throughput nucleic acid and absorbance assays with minimal sample consumption.
  • Contains one microplate adapter for automated plate handling.
  • Includes five 24-well low-volume bottom slides.
  • Includes five 64-well low-volume bottom slides.
  • Includes five 2 μL cover slides and five 4 μL cover slides.
  • Supports 2 μL and 4 μL sample volumes (approx. 0.5 mm and 1 mm pathlengths).
  • Suitable for high-throughput nucleic acid quantitation and absorbance measurements.
Catalog No. 50-000-14374 Supplier Molecular Devices Supplier No. 02007027
Add to Cart
Add to Cart
Request a Quote
The SpectraDrop High-Throughput Screening (HTS) Kit provides consumables and a microplate adapter to enable automated, low-volume sample measurement using micro-volume microplates. It combines multiple low-volume slide formats and cover slides to support high-throughput nucleic acid and absorbance assays with minimal sample consumption.
  • Contains one microplate adapter for automated plate handling.
  • Includes five 24-well low-volume bottom slides.
  • Includes five 64-well low-volume bottom slides.
  • Includes five 2 μL cover slides and five 4 μL cover slides.
  • Supports 2 μL and 4 μL sample volumes (approx. 0.5 mm and 1 mm pathlengths).
  • Suitable for high-throughput nucleic acid quantitation and absorbance measurements.