Learn More
Research Products International Corp Spectrafuge, 24D Micro-Centrifuge, Holds 24 x 1.5 ml Tubes, Blue
SDP
Catalog No. 50550358
Encompass
The ultimate micro-centrifuge designed for current laboratory applications. Powered by a maintenance free brushless motor that performs quietly without generating excessive heat. Motor design also provides quick acceleration and deceleration. Variable speed control allows precise selection. Ideal for many applications including microfiltration and separation of proteins and nucleic acids. Centrifuge can be set to run timed, continuous or quick spin. Angled rotor removes easily for cleaning. Supplied with a snap-on transparent cover that reduces noise levels. Unique ducting system provides a continuous air flow to keep the rotor cool and prevent elevation of sample temperatures.
Catalog No. 50-550-358 Supplier Research Products International Corp Supplier No. 141307B
Add to Cart
Add to Cart
The ultimate micro-centrifuge designed for current laboratory applications. Powered by a maintenance free brushless motor that performs quietly without generating excessive heat. Motor design also provides quick acceleration and deceleration. Variable speed control allows precise selection. Ideal for many applications including microfiltration and separation of proteins and nucleic acids. Centrifuge can be set to run timed, continuous or quick spin. Angled rotor removes easily for cleaning. Supplied with a snap-on transparent cover that reduces noise levels. Unique ducting system provides a continuous air flow to keep the rotor cool and prevent elevation of sample temperatures.