Learn More
Pramana SpectralHT-2 Tissue
Description
- Two (2) Spectral Scanners with Compute Units
- Robot Arm with the 240/load automated slide delivery
- Scanning Tables
- NAS Cluster Storage
- Network Switch and accessories.
- Magnification 40X, 0.8NA (0.25mpp)
Specifications
Specifications
| For Use With (Application) | Microarray Processing |