Learn More
Pramana SpectralHT-4 Tissue, Liquids, and Cells
Catalog No. 23888000
Change view
Click to view available options
For Use With (Application):
Microarray Processing
Catalog No. For Use With (Application)
23-888-000 Microarray Processing
1 options
Catalog No. 23-888-000 Supplier Pramana Supplier No. P4HTB480
Only null left
Add to Cart
Add to Cart
Request a Quote
Four (4) Head High Throughput System with Robotic Handler: Designed and Configured for Microbiology, Cytology, Bacteriology, and Blood Smears with the 480/load automated slide delivery.
  • Four (4) Spectral Scanners with Compute Units
  • Robot Arm with the 480/load automated slide delivery
  • Scanning Tables
  • NAS Cluster Storage
  • Network Switch and accessories.
  • Designed and Configured for Microbiology, Cytology, Bacteriology, and Blood Smears
  • Magnification 80X, 0.95NA (0.14mpp)
  • Magnification 40X, 0.8NA (0.25mpp)
Specifications
For Use With (Application) Microarray Processing