Learn More
Pramana SpectralHT-4 Tissue
Description
- Four (4) Spectral Scanners with Compute Unit
- Robot Arm with the 480/load automated slide delivery
- Scanning Tables
- NAS Cluster Storage
- Network Switch and accessories.
- Magnification 40X, 0.8NA (0.25mpp)
Specifications
Specifications
| For Use With (Application) | Microarray Processing |