Learn More
Pramana SpectralM-Pro Cassette
Catalog No. 23888028
Change view
Click to view available options
:
For Use With (Application): Microarray Processing
Catalog No.
23-888-028 For Use With (Application): Microarray Processing
1 options
Catalog No. 23-888-028 Supplier Pramana Supplier No. M1CSST4
Only null left
Add to Cart
Add to Cart
Desktop Scanner Manual Cassette: Four (4) slide capacity cassette supporting 1x3 slides.
  • Pramana Desktop Scanner Manual Cassette
  • Four (4) slide capacity cassette supporting 1x3 slides
  • Compatible with Pramana Desktop Scanner M1APRO or M1BPRO
  • M scanner Cassette 1x3, 4 Slide