Learn More
Pramana SpectralM-Pro Tissue, Liquids, and Cells
Catalog No. 23888008
Change view
Click to view available options
For Use With (Application):
Microarray Processing
Catalog No. For Use With (Application)
23-888-008 Microarray Processing
1 options
Catalog No. 23-888-008 Supplier Pramana Supplier No. M1BPRO
Add to Cart
Add to Cart
Desktop Scanner with Manual Cassette Loader: Designed and Configured for Microbiology, Cytology, Bacteriology, and Blood Smears with the four (4) slide manual cassette delivery.
  • Pramana Desktop Scanner with Manual Cassette Loader
  • Four (4) slide manual cassette delivery
  • Spectral-M-Pro scanner with Compute unit In-line Quality Assurance
  • Intelligent tissue locator
  • Volume Scanning
  • Image fusion
  • User control for scanning parameters
  • Integrated WSI Viewer
  • Quality Control Software
  • Integration with Pramana Cluster Management System (CMS)
  • Algorithm-builder and slide -loader utility for Edge AI
  • Designed and Configured for Microbiology, Cytology, Bacteriology, and Blood Smears
  • Magnification 80X, 0.95NA (0.14mpp)
  • Magnification 40X, 0.8NA (0.25mpp)
Specifications
For Use With (Application) Microarray Processing