Learn More
Pramana SpectralM-Pro Tissue
Description
- Pramana Desktop Scanner with Manual Cassette Loader
- Four (4) slide manual cassette delivery
- Spectral-M-Pro scanner with Compute unit In-line Quality Assurance
- Intelligent tissue locator
- Volume Scanning
- Image fusion
- User control for scanning parameters
- Integrated WSI Viewer
- Quality Control Software
- Integration with Pramana Cluster Management System (CMS)
- Magnification 40X to 0.8NA (0.25mpp)
Specifications
Specifications
| For Use With (Application) | Microarray Processing |