Learn More
Med Vet International Spectramast Dc Ceftiofur Hcl Syringes, 500mg/10mL, 12/Box
Catalog No. 502828644
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Ceftiofur HCl sterile suspension for intramammary infusion in dry dairy cattle only. Each 10-mL PLASTET disposable syringe contains 500 mg ceftiofur equivalents as the hydrochloride salt.
Catalog No. 50-282-8644 Supplier Med Vet International Supplier No. RXSPECTRADC
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Ceftiofur HCl sterile suspension for intramammary infusion in dry dairy cattle only. Each 10-mL PLASTET disposable syringe contains 500 mg ceftiofur equivalents as the hydrochloride salt.