Learn More
Med Vet International Spectramast Lc (Ceftiofur Hydrochloride) Sterile Suspension, Lactating Cow Formula, 10mL, 144 Syringes
Catalog No. 502828875
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
SPECTRAMAST LC (ceftiofur intramammary suspension) Sterile Suspension is indicated for use in lactating dairy cattle for (1) the treatment of clinical mastitis associated with coagulase-negative staphylococci, Streptococcus dysgalactiae, and Escherichia coli and (2) the treatment of diagnosed subclinical mastitis associated with coagulase-negative staphylococci and Streptococcus dysgalactiae and (3) help treat diagnosed subclinical mastitis infections before lactating cows exhibit physical symptoms of the disease.
Catalog No. 50-282-8875 Supplier Med Vet International Supplier No. RXSPECTRAMASTLCP
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
SPECTRAMAST LC (ceftiofur intramammary suspension) Sterile Suspension is indicated for use in lactating dairy cattle for (1) the treatment of clinical mastitis associated with coagulase-negative staphylococci, Streptococcus dysgalactiae, and Escherichia coli and (2) the treatment of diagnosed subclinical mastitis associated with coagulase-negative staphylococci and Streptococcus dysgalactiae and (3) help treat diagnosed subclinical mastitis infections before lactating cows exhibit physical symptoms of the disease.