Learn More
Molecular Devices Molecular Devices Absorbance Microplate Reader, 384-well, 190-1000 nm, Temperature Controlled
Catalog No. 5000014731
Encompass_Preferred
The SpectraMax Plus 384 absorbance microplate reader from Molecular Devices provides UV-visible absorbance measurements for 96- and 384-well plates, as well as standard cuvettes and test tubes. It combines full spectral scanning, multiple read modes, and plate temperature control to support routine assays and higher-throughput workflows.
  • Full spectral scanning from 190 to 1000 nm for UV-visible measurements.
  • Supports 96-well, 384-well, and strip-well plates, plus standard cuvettes and 12 x 75 mm test tubes.
  • Endpoint, kinetic, and spectrum read modes for diverse assay formats.
  • Pathlength correction normalizes absorbance to a 1 cm path length.
  • Plate chamber temperature control from 4°C above ambient to 45°C with < 1.0°C variation.
  • Dedicated cuvette chamber with percent transmission or absorbance display and reference correction.
  • Designed for both routine 96-well workflows and high-throughput 384-well assays.
Catalog No. 50-000-14731 Supplier Molecular Devices Supplier No. PLUS384G
Add to Cart
Add to Cart
Request a Quote
The SpectraMax Plus 384 absorbance microplate reader from Molecular Devices provides UV-visible absorbance measurements for 96- and 384-well plates, as well as standard cuvettes and test tubes. It combines full spectral scanning, multiple read modes, and plate temperature control to support routine assays and higher-throughput workflows.
  • Full spectral scanning from 190 to 1000 nm for UV-visible measurements.
  • Supports 96-well, 384-well, and strip-well plates, plus standard cuvettes and 12 x 75 mm test tubes.
  • Endpoint, kinetic, and spectrum read modes for diverse assay formats.
  • Pathlength correction normalizes absorbance to a 1 cm path length.
  • Plate chamber temperature control from 4°C above ambient to 45°C with < 1.0°C variation.
  • Dedicated cuvette chamber with percent transmission or absorbance display and reference correction.
  • Designed for both routine 96-well workflows and high-throughput 384-well assays.