Learn More
Molecular Devices Molecular Devices SpectraMax i3x Microplate Reader, Multi-Mode, 6-1536 Well
Catalog No. 5000014696
Encompass_Preferred
The SpectraMax i3x is a configurable multi-mode microplate reader designed for biochemical and cell-based assays. It provides absorbance, fluorescence, and luminescence detection across a broad wavelength range, supports 6-1536 well plates, and can be field-upgraded with imaging, TRF/HTRF, AlphaScreen, injectors, and western blot detection modules.
  • Configurable multi-mode detection for absorbance, fluorescence, and luminescence.
  • Supports plate formats from 6 to 1536 wells.
  • Broad wavelength coverage from UV to near-infrared.
  • Field-upgradeable modules for imaging, TRF/HTRF, AlphaScreen, injectors, and western blot detection.
  • High-sensitivity optics with xenon flash lamp and LED illumination options.
  • Compact footprint suitable for bench or automated workflows.
Catalog No. 50-000-14696 Supplier Molecular Devices Supplier No. I3XG
Add to Cart
Add to Cart
Request a Quote
The SpectraMax i3x is a configurable multi-mode microplate reader designed for biochemical and cell-based assays. It provides absorbance, fluorescence, and luminescence detection across a broad wavelength range, supports 6-1536 well plates, and can be field-upgraded with imaging, TRF/HTRF, AlphaScreen, injectors, and western blot detection modules.
  • Configurable multi-mode detection for absorbance, fluorescence, and luminescence.
  • Supports plate formats from 6 to 1536 wells.
  • Broad wavelength coverage from UV to near-infrared.
  • Field-upgradeable modules for imaging, TRF/HTRF, AlphaScreen, injectors, and western blot detection.
  • High-sensitivity optics with xenon flash lamp and LED illumination options.
  • Compact footprint suitable for bench or automated workflows.