Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, Dual Monochromators, 6-384-Well
Catalog No. 5000014352
Encompass_Preferred
The SpectraMax M5e is a modular multi-mode microplate reader designed for a wide range of life science assays. It provides flexible wavelength selection and sensitive detection across multiple modes while supporting automation for increased throughput.
  • Performs absorbance, fluorescence intensity, luminescence, time-resolved fluorescence, and fluorescence polarization measurements.
  • Dual scanning monochromators with 1 nm spectral scanning for flexible excitation and emission selection.
  • Spectral ranges: absorbance 200-1000 nm; fluorescence/luminescence/TRF 250-850 nm; fluorescence polarization 300-750 nm.
  • Supports 6-384 well microplates and cuvette-based measurements.
  • Compatible with automated plate handling, including a StakMax stacker with up to 50-plate capacity and optional barcode reader.
  • Includes PathCheck sensor for absorbance normalization and AutoPMT optimization for extended dynamic range.
Catalog No. 50-000-14352 Supplier Molecular Devices Supplier No. M5E
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M5e is a modular multi-mode microplate reader designed for a wide range of life science assays. It provides flexible wavelength selection and sensitive detection across multiple modes while supporting automation for increased throughput.
  • Performs absorbance, fluorescence intensity, luminescence, time-resolved fluorescence, and fluorescence polarization measurements.
  • Dual scanning monochromators with 1 nm spectral scanning for flexible excitation and emission selection.
  • Spectral ranges: absorbance 200-1000 nm; fluorescence/luminescence/TRF 250-850 nm; fluorescence polarization 300-750 nm.
  • Supports 6-384 well microplates and cuvette-based measurements.
  • Compatible with automated plate handling, including a StakMax stacker with up to 50-plate capacity and optional barcode reader.
  • Includes PathCheck sensor for absorbance normalization and AutoPMT optimization for extended dynamic range.