Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, Monochromator, Temperature Control
Catalog No. 5000014721
Encompass_Preferred
The SpectraMax M5e multi-mode microplate reader provides flexible, high-performance detection for absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF assays across 6-384 well plates and cuvettes. It features a tunable monochromator with 1 nm spectral resolution, temperature control, and adaptive detector optimization for broad dynamic range.
  • Supports absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF assays.
  • Monochromator with 1 nm wavelength selection for spectral scanning and up to six wavelengths per read.
  • Temperature control from ambient +2°C to 60°C for incubated assays.
  • Automated detector optimization and absorbance normalization for extended dynamic range and improved accuracy.
  • Compatible with GxP-capable data acquisition and validation workflows for regulated environments.
Catalog No. 50-000-14721 Supplier Molecular Devices Supplier No. M5EG
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M5e multi-mode microplate reader provides flexible, high-performance detection for absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF assays across 6-384 well plates and cuvettes. It features a tunable monochromator with 1 nm spectral resolution, temperature control, and adaptive detector optimization for broad dynamic range.
  • Supports absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF assays.
  • Monochromator with 1 nm wavelength selection for spectral scanning and up to six wavelengths per read.
  • Temperature control from ambient +2°C to 60°C for incubated assays.
  • Automated detector optimization and absorbance normalization for extended dynamic range and improved accuracy.
  • Compatible with GxP-capable data acquisition and validation workflows for regulated environments.