Learn More
Molecular Devices Molecular Devices Desktop Computer, SpectraMax MiniMax 300, USB 3.0
Catalog No. 5000014670
Encompass_Preferred
The desktop computer supplied for the SpectraMax MiniMax 300 imaging cytometer is a tested, preconfigured PC that provides the required ports, software, and system settings for reliable instrument control, image acquisition, and data handling. The computer is shipped separately from the monitor and includes user-recommended power and OS settings to prevent data loss.
  • Preconfigured for instrument control and image acquisition
  • Dedicated USB 3.0 ports for camera connection
  • HDMI output with DVI cable and HDMI-to-DVI adapter for monitor connection
  • Preinstalled instrument software and productivity software (activation required)
  • Power-management settings set to prevent sleep, hibernate, or hard-disk spin-down
  • Supplied connection cables included for instrument integration
Catalog No. 50-000-14670 Supplier Molecular Devices Supplier No. MINIMAXCOMPUTER
Add to Cart
Add to Cart
Request a Quote
The desktop computer supplied for the SpectraMax MiniMax 300 imaging cytometer is a tested, preconfigured PC that provides the required ports, software, and system settings for reliable instrument control, image acquisition, and data handling. The computer is shipped separately from the monitor and includes user-recommended power and OS settings to prevent data loss.
  • Preconfigured for instrument control and image acquisition
  • Dedicated USB 3.0 ports for camera connection
  • HDMI output with DVI cable and HDMI-to-DVI adapter for monitor connection
  • Preinstalled instrument software and productivity software (activation required)
  • Power-management settings set to prevent sleep, hibernate, or hard-disk spin-down
  • Supplied connection cables included for instrument integration