Learn More
Molecular Devices Molecular Devices Automated Cell Imaging System, Digital Confocal 2D, 6-384-Well
Catalog No. 5000014676
Encompass_Preferred
The ImageXpress Pico Advanced is a compact benchtop automated cell imaging system that provides fluorescence and brightfield imaging with on-the-fly digital confocal deconvolution and integrated analysis for 6-384 well assays and live-cell experiments.
  • Automated fluorescence and brightfield imaging with digital confocal 2D deconvolution.
  • 6-position automated turret with user-exchangeable high-NA air objectives.
  • Supports 6-384-well microplates, slides, and 35 mm dishes.
  • Autofocus, live preview, z-stack acquisition, and robotic plate handler compatibility.
  • Includes icon-driven acquisition and analysis software with preconfigured assay templates.
  • Optional environmental control for temperature, CO2, O2, and humidity for live-cell work.
Catalog No. 50-000-14676 Supplier Molecular Devices Supplier No. PICOADV
Add to Cart
Add to Cart
Request a Quote
The ImageXpress Pico Advanced is a compact benchtop automated cell imaging system that provides fluorescence and brightfield imaging with on-the-fly digital confocal deconvolution and integrated analysis for 6-384 well assays and live-cell experiments.
  • Automated fluorescence and brightfield imaging with digital confocal 2D deconvolution.
  • 6-position automated turret with user-exchangeable high-NA air objectives.
  • Supports 6-384-well microplates, slides, and 35 mm dishes.
  • Autofocus, live preview, z-stack acquisition, and robotic plate handler compatibility.
  • Includes icon-driven acquisition and analysis software with preconfigured assay templates.
  • Optional environmental control for temperature, CO2, O2, and humidity for live-cell work.