Learn More
Research Products International Corp SpectraTube Rainbow Cap Centrifuge Tubes, Red, 50ml Graduated, Bulk, 25 per Bag, 500 per Case
SDP
Catalog No. 501367526
Encompass
SpectraTubes feature colored caps for easy sample identification. Caps are brightly colored, frosted and easy to write on with any marking pen. Conical tubes are made from high clarity polypropylene which features excellent chemical and temperature resistance. Tubes withstand temperature extremes from -45°C to +120°C.
Tubes are supplied in bulk pack bags for convenience. A case of 500 tubes contains bags of 25 tubes each in 5 colors: Red, Yellow, Blue, Orange and Green.
Catalog No. 50-136-7526 Supplier Research Products International Corp Supplier No. 151066
SpectraTubes feature colored caps for easy sample identification. Caps are brightly colored, frosted and easy to write on with any marking pen. Conical tubes are made from high clarity polypropylene which features excellent chemical and temperature resistance. Tubes withstand temperature extremes from -45°C to +120°C.
Tubes are supplied in bulk pack bags for convenience. A case of 500 tubes contains bags of 25 tubes each in 5 colors: Red, Yellow, Blue, Orange and Green.