Learn More
Research Products International Corp SpectraTube Rainbow Cap Centrifuge Tubes, 50ml Graduated, 25 Tubes per Tray, 500 Tubes per Case
SDP
Catalog No. 501367525
Encompass
SpectraTubes feature colored caps for easy sample identification. Caps are brightly colored, frosted and easy to write on with any marking pen. Conical tubes are made from high clarity polypropylene which features excellent chemical and temperature resistance. Tubes withstand temperature extremes from -45°C to +120°C.
Tubes are supplied in trays for quick retrieval.
Catalog No. 50-136-7525 Supplier Research Products International Corp Supplier No. 151065
SpectraTubes feature colored caps for easy sample identification. Caps are brightly colored, frosted and easy to write on with any marking pen. Conical tubes are made from high clarity polypropylene which features excellent chemical and temperature resistance. Tubes withstand temperature extremes from -45°C to +120°C.
Tubes are supplied in trays for quick retrieval.