Learn More
Precision Glassblowing of Colorado SPECTRO CHAMBER 48105061
SDP
Catalog No. 50420758
Encompass_Preferred
Modified Lichte Cyclonic Spray Chamber, Pump Drain, for Spectro Modula, CIROS, Genesis, & ARCOS
Catalog No. 50-420-758 Supplier Precision Glassblowing of Colorado Supplier No. PG51027
Modified Lichte Cyclonic Spray Chamber, Pump Drain, for Spectro Modula, CIROS, Genesis, & ARCOS