Learn More
Research Products International Corp Spectro Cuvette Rack, Poly, 20x10mm Cells
SDP
Catalog No. 501978346
Encompass
This polypropylene rack holds twenty 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells. Wells hold standard cuvettes with a 12.5mm cross-section. Sloped top makes the second tier more accessible. Steam autoclavable at 121ºC (250ºF).
Catalog No. 50-197-8346 Supplier Research Products International Corp Supplier No. 249003
This polypropylene rack holds twenty 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells. Wells hold standard cuvettes with a 12.5mm cross-section. Sloped top makes the second tier more accessible. Steam autoclavable at 121ºC (250ºF).