Learn More
Precision Glassblowing of Colorado SPECTRO NEBULIZER 76060529
SDP
Catalog No. 50420757
Encompass_Preferred
Modified Lichte MDSN Nebulizer, 1 lpm Ar, with Spectro Fittings
Catalog No. 50-420-757 Supplier Precision Glassblowing of Colorado Supplier No. PG51020SF
Modified Lichte MDSN Nebulizer, 1 lpm Ar, with Spectro Fittings