Learn More
Ted Pella Inc Spectro Tabs, High Purity Conductive Carbon Tabs, 12mm O.D., 120/pk
SDP
Catalog No. NC9967922
Encompass
Spectro brand tabs have been a standard for quality carbon tabs for many years. Construction and thickness is similar to the PELCO Tabs™ Carbon Conductive Tabs above. The EDX Analysis shows only minor impurities of Al, Si, and some localized sulfur particles may be found in the adhesive.
Catalog No. NC9967922 Supplier Ted Pella Inc Supplier No. 160844
Spectro brand tabs have been a standard for quality carbon tabs for many years. Construction and thickness is similar to the PELCO Tabs™ Carbon Conductive Tabs above. The EDX Analysis shows only minor impurities of Al, Si, and some localized sulfur particles may be found in the adhesive.