Learn More
Thermo Scientific™ Spectroflux 128, Lithium tetraborate, Lithium metaborate & Sodium nitrate, 33.0:61.5:5.5 w/w%
Catalog No. AA12090A1
Change view
Click to view available options
Quantity:
1 kg
Catalog No. Quantity
AA12090A1 1 kg
1 options
Catalog No. AA12090A1 Supplier Thermo Scientific™ Supplier No. 12090A1