Learn More
Thermo Scientific™ Spectroflux 135, Lithium tetraborate, Lithium fluoride, Strontium oxide & Vanadium(V) oxide, 84.9:8.4:4.6:2.1 w/w%
Catalog No. AA12091A1
Change view
Click to view available options
Quantity:
1 kg
Catalog No. Quantity
AA12091A1 1 kg
1 options
Catalog No. AA12091A1 Supplier Thermo Scientific™ Supplier No. 12091A1