Learn More
Spectroline™ ImageAide Autoanalysis Software
SDP
Catalog No. 11992349 Shop All Spectronics Corporation Products
Change view
Click to view available options
Software, Upgrade,imageaide,ea
Software,analysis,imageaide,ea
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-992-349 Software, Upgrade,imageaide,ea
11-992-351 Software,analysis,imageaide,ea
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-992-349 Supplier Spectroline™ Supplier No. GL2500
Includes
Acquisition Board, CCD Camera, cables, and software
  • For customizable analysis of digital images of all gel media, colonies, spots, and blots
  • Imports camera images and automatically finds lanes, bands, and edges, producing track profiles and results in less than ten seconds
  • Determines molecular weights, quantity, and Rf, then displays all in a single window; also allows data export directly into an Excel™ spreadsheet and Microsoft™ Word™ document
  • Autotrack finding feature detects distorted or misshapen tracks and bands
  • Supplied with the Spectroline™ GeneLine, ImageEZE, DigiCAM, or Epi-DigiCAM series; available also as separate analysis software package