Learn More
Chemplex Industries SPECTROPELLET FILM 32MM
Catalog No. 50249376
Encompass_Preferred
CHEMPLEX SpectroPellet(TM) Protective Die Film for 32 mm dies, 500 circles/pkg
Catalog No. 50-249-376 Supplier Chemplex Industries Supplier No. 7032
CHEMPLEX SpectroPellet(TM) Protective Die Film for 32 mm dies, 500 circles/pkg