Learn More
Starna Cells Inc SPECTROPHOMETER CELL/W SEALCAP
SDP
Catalog No. NC9929386
Encompass_Preferred
Cell with Sealing cap Path Length: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 68mm Interior Width: 10mm Interior Length: 10mm Nominal Volume: 3.500mL Number of Polished Windows: 2 useable range: 170 to 2700nm
Catalog No. NC9929386 Supplier Starna Cells Inc Supplier No. 1Q10GL14C
Cell with Sealing cap Path Length: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 68mm Interior Width: 10mm Interior Length: 10mm Nominal Volume: 3.500mL Number of Polished Windows: 2 useable range: 170 to 2700nm